DL-Leucin

Lieferant: BACHEM BIOCHEMICA

Synonyms: (±)-Leucin(±)-2-Amino-4-methylpentansäure(±)-α-Amino-γ-methylvaleriansäureDL-α-Amino-γ-methylvaleriansäure

BACE4030793.0100EA 0 CHF
BACE4030793.0100 BACE4030793.0025
DL-Leucin
DL-Leucin
Formel: C₆H₁₃NO₂
MW: 131,17 g/mol
Siedepunkt: 226 °C (1013 hPa)
Dichte: 1,191 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00063087
CAS-Nummer: 328-39-2
EINECS: 206-328-2
Merck Index: 13,05470

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR