Dibenzyldiselenid

Lieferant: Molekula
Gefahr

MOLEM54807763EA 0 CHF
MOLEM54807763
Dibenzyldiselenid
Dibenzyldiselenid
Formel: C₁₄H₁₄Se₂
MW: 340,19 g/mol
Melting Pt: 91…94 °C
Dichte: 1,67 g/cm³ (25 °C)
MDL Number: MFCD00004767
CAS-Nummer: 1482-82-2
UN: 3283
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR