Anti-Aromatase Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BOSSBS-1292R-A680EA 1150 CHF
BOSSBS-1292R-A680
Anti-Aromatase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyse many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localises to the endoplasmic reticulum and catalyses the last steps of estrogen biosynthesis, three successive hydroxylations of the A ring of androgens. Mutations in this gene can result in either increased or decreased aromatase activity; the associated phenotypes suggest that estrogen functions both as a sex steroid hormone and in growth or differentiation. The gene expresses two transcript variants. Belongs to the cytochrome P450 family.

Type: Primary
Antigen: Aromatase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-140/503
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR