Anti-CASP3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BOSSBS-0081R-A750EA 1150 CHF
BOSSBS-0081R-A750
Anti-CASP3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Involved in the activation cascade of caspases responsible for apoptosis execution. At the onset of apoptosis it proteolytically cleaves poly(ADP-ribose) polymerase (PARP) at a '216-Asp-|-Gly-217' bond. Cleaves and activates sterol regulatory element binding proteins (SREBPs) between the basic helix-loop-helix leucine zipper domain and the membrane attachment domain. Cleaves and activates caspase-6, -7 and -9. Involved in the cleavage of huntingtin. Triggers cell adhesion in sympathetic neurons through RET cleavage.

Type: Primary
Antigen: Caspase-3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/277
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Pig, Chicken, Sheep, Rabbit, Others, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR