Anti-MTNR1A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BOSSBS-0027R-A750EA 1150 CHF
BOSSBS-0027R-A750
Anti-MTNR1A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.
Type: Primary
Antigen: Melatonin Receptor 1A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-280/350
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Others