Anti-DNA polymerase mu Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BOSSBS-13021R-A680EA 1140 CHF
BOSSBS-13021R-A680
Anti-DNA polymerase mu Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
DNA polymerase mu shares a number of characteristics with DNA polymerase Beta as well as with terminal deoxynucleotideyltransferase. Pol mu purportedly plays a role in microhomology mediated joining and the repair of double-stranded breaks. However, unlike other DNA polymerases, which show substrate specificity for deoxynucleotides, DNA Pol mu incorporates both deoxynucleotides and ribonucleotides in a template- directed manner. This unusual capability implies a novel role for this polymerase in DNA repair.

Type: Primary
Antigen: DNA polymerase mu
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 261-360/494
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR