(S)-(+)-2-Hydroxyisovaleriansäure 97%

Lieferant: Apollo Scientific
Achtung

APOSOR17815-10GEA 38.5 CHF
APOSOR17815-10G APOSOR17815-25G
(S)-(+)-2-Hydroxyisovaleriansäure 97%
(S)-(+)-2-Hydroxyisovaleriansäure
Formel: C₅H₁₀O₃
MW: 118,13 g/mol
Siedepunkt: 124…125 °C/13mmHg
Melting Pt: 68…70 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00066443
CAS-Nummer: 17407-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR