Methyloleat

Lieferant: MATREYA

Synonyms: Ölsäuremethylestercis-9-Octadecensäuremethylester

MAYA1023EA 0 CHF
MAYA1023 BTRN1023
Methyloleat
Methyloleat
Formel: C₁₉H₃₆O₂
MW: 296,49 g/mol
Siedepunkt: ∼218,5 °C (1013 hPa)
Melting Pt: –20 °C
Dichte: 0,88 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009578
CAS-Nummer: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR