L(+)-allo-Isoleucin

Lieferant: Molekula

MOLE14645122-250MGEA 0 CHF
MOLE14645122-250MG
L(+)-allo-Isoleucin
L(+)-allo-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131,17 g/mol
MDL Number: MFCD00066446
CAS-Nummer: 1509-34-8
EINECS: 216-142-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR