Anti-FUT4 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BOSSBS-1702R-CY3EA 1160 CHF
BOSSBS-1702R-CY3
Anti-FUT4 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
May catalyze alpha-1,3 glycosidic linkages involved in the expression of Lewis X/SSEA-1 and VIM-2 antigens.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: CD15/Fut4/SSEA-1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR