Anti-MTNR1A Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BOSSBS-0027R-CY3EA 1160 CHF
BOSSBS-0027R-CY3
Anti-MTNR1A Rabbit Polyclonal Antibody (Cy3®)
Antikörper
High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: MTNR1A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR