Anti-PPARA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK600-401-421SEA 254 CHF
ROCK600-401-421S
Anti-PPARA Rabbit Polyclonal Antibody
Antikörper
This affinity purified antibody has been tested for use in ELISA and by western blot. Expect a single band approximately 52 kDa in size corresponding to PPAR alpha by western blot in the appropriate tissue or cell lysate.

Recommended Dilutions: ELISA: 1:8000 - 1:32000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:100-1:300;

Type: Primary
Antigen: PPARA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Bovine, Dog, Golden Hamster, Boar, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR