Anti-Protein A Goat Polyclonal Antibody (Peroxidase)
Lieferant: Rockland Immunochemicals
ROCK200-1377EA 6470 CHF
ROCK200-1377
Anti-Protein A Goat Polyclonal Antibody (Peroxidase)
Antikörper
Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope:
Host: Goat
Isotype: none
Reactivity: Pan