Anti-HSPA1A Rabbit Polyclonal Antibody

Lieferant: Bioss
BOSSBS-0126REA 955 CHF
BOSSBS-0126R
Anti-HSPA1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSPA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR