1,2-Benzisothiazol-3(2H)-on

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS20336G100EA 120 CHF
ACTIAS20336G100 ACTIAS20336G25
1,2-Benzisothiazol-3(2H)-on
1,2-Benzisothiazol-3(2H)-on
Formel: C₇H₅NOS
MW: 151,19 g/mol
MDL Number: MFCD00044001
CAS-Nummer: 2634-33-5
EINECS: 220-120-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR