D-allo-Threonin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS33080G5EA
0
CHF
ACTIAS33080G5
ACTIAS33080G1
D-allo-Threonin
D-allo-Threonin
Formel:
C₄H₉NO₃ MW: 119,12 g/mol |
MDL Number:
MFCD00004526 CAS-Nummer: 24830-94-2 EINECS: 246-488-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...