4'-Amino-2'-methylacetanilid
Lieferant: ACTIVATE SCIENTIFIC
AS1650G1
AS1650G5
ACTIAS1650G1EA
0
CHF
ACTIAS1650G1
ACTIAS1650G5
4'-Amino-2'-methylacetanilid
4'-Amino-2'-methylacetanilid
Formel:
C₉H₁₂N₂O |
MDL Number:
MFCD00276633 CAS-Nummer: 56891-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...