Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 488)

Lieferant: Rockland Immunochemicals
612-141-120
ROCK612-141-120EA 256 CHF
ROCK612-141-120
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 488)
Antikörper
The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR