Ricinoleic acid ≥99%, Flüssigkeit

Lieferant: ICN Biomedicals

Synonyms: Ricinolic acid, (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid, Ricinelaidic acid, (R,Z)-12-Hydroxyoctadec-9-enoic acid

0219151490 0219151480
ICNA0219151490EA 431 CHF
ICNA0219151490 ICNA0219151480
Ricinoleic acid ≥99%, Flüssigkeit
Ricinoleic acid

Ricinoleic Acid is a naturally occurring 12-hydroxy fatty acid constituting about 90% of the fatty acids in castor oil. Has the capacity to serve as a substrate for the synthesis of the conjugated linoleic acids.


Ricinoleic Acid is used as a substrate for the synthesis of the conjugated linoleic acids.

Formel: C₁₈H₃₄O₃
MDL Number: MFCD00084840
CAS-Nummer: 141-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR