2-Hydroxy-1-naphthaldehyd

Lieferant: ICN Biomedicals

Synonyms: 2-Hydroxy-α-naphthaldehyde

0215129825
ICNA0215129825EA 34 CHF
ICNA0215129825
2-Hydroxy-1-naphthaldehyd
2-Hydroxy-1-naphthaldehyd
Formel: C₁₁H₈O₂
MDL Number: MFCD00004005
CAS-Nummer: 708-06-5
EINECS: 211-902-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR