n-Nonadecan

Lieferant: EHRENSTORFER

Synonyms: N-nonadecane

C15622000
EHERC15622000EA 65.4 CHF
EHERC15622000
n-Nonadecan
n-Nonadecan
Formel: H₃C(CH₂)₁₇CH₃
Siedepunkt: 329.7 °C (1013 hPa)
Melting Pt: 32.5 °C
Dichte: 0.7845 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009012
CAS-Nummer: 629-92-5
EINECS: 211-116-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR