Diphenylsulfon

Lieferant: EHRENSTORFER

Synonyms: Phenyl sulphone, sulphonyldibenzene

C12915000
EHERC12915000EA 75.8 CHF
EHERC12915000
Diphenylsulfon
Diphenylsulfon
Formel: C₁₂H₁₀O₂S
MDL Number: MFCD00007548
CAS-Nummer: 127-63-9
EINECS: 204-853-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR