Anti-Met-Enkephalin Rabbit Polyclonal Antibody
Lieferant: Biosensis
R-1499-50
BSENR-1499-50EA 374 CHF
BSENR-1499-50
Anti-Met-Enkephalin Rabbit Polyclonal Antibody
Antikörper
Human Methionine enkephalin (Met-Enkephalin) is a small 5 amino acid peptide cleaved from the precursor pro-opiomelanocortin (POMC). Met-Enkephalin is an endogenous opioid peptide that interacts with opioid receptors and produces analgesic effects.
Antibody recognizes Met-Enkephalin in native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrationsMore Product Information