Ammoniaklösung 25% in Wasser zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

390030010 390030025
ACRO390030010EA 47.4 CHF
ACRO390030010 ACRO390030025
Ammoniaklösung 25% in Wasser zur Analyse
Ammoniaklösung
Formel: NH₃
Siedepunkt: 38 °C (760 mmHg)
Melting Pt: –57 °C
Dichte: 0.91 g/cm³
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011418
CAS-Nummer: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless Solution
Non-volatile matter ≤0.002 %
Titration with HCl ≥25 % (as NH3)
Chloride (Cl) ≤0.00005 %
Sulfide (S) ≤0.0002 %
Carbonate (CO3) ≤0.001 %
Phosphate (PO4) ≤0.00005 %
Sulfate (SO4) ≤0.0002 %
Barium (Ba) ≤0.00001 %
Bismuth (Bi) ≤0.00001 %
Cadmium (Cd) ≤0.000005 %
Calcium (Ca) ≤0.0001 %
Chromium (Cr) ≤0.000005 %
Cobalt (Co) ≤0.00001 %
Copper (Cu) ≤0.00001 %
Gallium (Ga) ≤0.000002 %
Indium (In) ≤0.000002 %
Iron (Fe) ≤0.00001 %
Lead (Pb) ≤0.000005 %
Lithium (Li) ≤0.000002 %
Magnesium (Mg) ≤0.00005 %
Manganese (Mn) ≤0.00001 %
Molybdenum (Mo) ≤0.00001 %
Nickel (Ni) ≤0.000005 %
Platinum (Pt) ≤0.00001 %
Potassium (K) ≤0.0001 %
Sodium (Na) ≤0.0001 %
Strontium (Sr) ≤0.0001 %
Thallium (Tl) ≤0.000005 %
Tin (Sn) ≤0.00001 %
Titanium (Ti) ≤0.00001 %
Zinc (Zn) ≤0.00001 %
Reducing substances ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR