2-Amino-2-methylpropanol 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP

104060010 104065000 104060050
ACRO104060010EA 84.5 CHF
ACRO104060010 ACRO104065000 ACRO104060050
2-Amino-2-methylpropanol 99%
AMP (2-Amino-2-methylpropanol)
Formel: (CH₃)₂C(NH₂)CH₂OH
Siedepunkt: 165 °C (1013 hPa)
Melting Pt: 31…32 °C
Dichte: 0.93 g/cm³
Flash-Pt: 67 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008051
CAS-Nummer: 124-68-5
EINECS: 204-709-8
Merck Index: 13,00448

Order Now

Spezifikation Testergebnisse

Appearance Colorless Low melting solid
Infrared spectrum Conforms
Water ≤0.8 %
Titration with HClO4 ≥98.5 %
UV at 420 nm A: ≤0.005
Color scale ≤10 APHA
Suitability (in liquid scint. counting) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR