Hotplates
Lieferant: NICKEL ELECTRO
HP1-1 HP1-2 HP1-3
453-0004EA 2280 CHF
453-0004 453-0006 453-0008
Hotplates
Heizgeräte Heizplatten
Stainless steel casing, stove enamelled finish and cast aluminium plate. Units accept retort rods.
  • Temperature controlled by hydraulic thermostat
  • With illuminated on/off switch and heater “ON” indicator
  • Fitted with non-slip rubber feet for safety
Each hotplate accepts an optional sand bath attachment with aluminium upstand rim which fits over edge of hotplate. The attachment can also act as a safety rim to prevent dishes falling off hotplate.
Order Now
TECHNISCHE DATEN
Environmentally Preferable